Orders LinkContactUsLinkComponent
2,4-Dinitrobenzaldehyde ≥98%
2,4-Dinitrobenzaldehyde ≥98%
Glass bottle for solids, 25 g
  A15452.14
2,4-Dinitrobenzaldehyde ≥98%
  A15452.14
 :  528-75-6
2,4-Dinitrobenzaldehyde ≥98%
2,4-Dinitrobenzaldehyde ≥98%
Glass bottle for solids, 25 g
 :  528-75-6
359,00 €
1 * 25 g
Purity: 98% CAS No.: 528-75-6 Molecular Formula: C7H4N2O5 Formula Weight: 196.12 Melting Point: 65-68° Boiling Point: 190°/10mm Merck Reference: 14,3272 MDL No.: MFCD00013376
  •  :528-75-6
  •  :C₇H₄N₂O₅
  •  :MFCD00013376
  •  :196.12 g/mol
  •  :14,3272
  •  :190 °C (10 mmHg)
  •  :65…68 °C
  •  :Ambient
  •  :208-440-7
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g