ω-Bromo-4-fluoroacetophenone ≥97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
ω-Bromo-4-fluoroacetophenone ≥97%
Catalog # A15319.06
Supplier: Thermo Fisher Scientific
CAS Number: 403-29-2
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 403-29-2 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 150°/12mm Beilstein Reference: 637863 MDL No.: MFCD00040830
Synonyms: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
- CAS Number:403-29-2
- Formula:C₈H₆BrFO
- MDL Number:MFCD00040830
- Molecular Weight:217.04 g/mol
- Boiling Pt:150 °C (12 mmHg)
- Melting Pt:45…49 °C
- Flash Pt:>110 °C (230 °F)
- EINECS:206-955-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



