Orders LinkContactUsLinkComponent
1,3-Propanedithiol ≥97%
Glass bottle for solids, 100 g
  A15261.22
1,3-Propanedithiol ≥97%
  A15261.22
 :  109-80-8
1,3-Propanedithiol ≥97%
1,3-Propanedithiol ≥97%
Glass bottle for solids, 100 g
 :  109-80-8
217,00 €
1 * 100 g
Purity: 97% CAS No.: 109-80-8 Molecular Formula: HS(CH2)3SH Formula Weight: 108.23 Flash Point: 63°(145°F) Refractive Index: 1.5400 Odour: Stench Melting Point: -79° Boiling Point: 168-170° Solubility: Slightly soluble in water. Miscible with alcohol, ether, chloroform, ether Beilstein Reference: 1071197 Merck Reference: 14,7801 UN No.: 3334 MDL No.: MFCD00004904
 :  1,3-Dimercaptopropane
  •  :109-80-8
  •  :C₃H₈S₂
  •  :MFCD00004904
  •  :108.23 g/mol
  •  :14,7801
  •  :168…170 °C
  •  :–79 °C
  •  :1.077
  •  :63 °C (145 °F)
  •  :Ambient
  •  :203-706-9
  •  :3334
  •  :9,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g