Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dimethoxybenzaldehyde ≥98%

3,5-Dimethoxybenzaldehyde ≥98%

Glass bottle for solids, 25 g
Catalog # A15109.14
Irritant
3,5-Dimethoxybenzaldehyde ≥98%
Catalog # A15109.14
CAS Number:  7311-34-4
3,5-Dimethoxybenzaldehyde ≥98%
3,5-Dimethoxybenzaldehyde ≥98%

Glass bottle for solids, 25 g

CAS Number:  7311-34-4
Price
200,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 7311-34-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 44-48° Boiling Point: 115-118°/1mm Beilstein Reference: 2042374 MDL No.: MFCD00003366

  • CAS Number:7311-34-4
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00003366
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:115…118 °C (1 mmHg)
  • Melting Pt:44…48 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:230-772-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications