3,5-Dimethoxybenzaldehyde ≥98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
3,5-Dimethoxybenzaldehyde ≥98%
Catalog # A15109.06
Supplier: Thermo Fisher Scientific
CAS Number: 7311-34-4
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 7311-34-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 44-48° Boiling Point: 115-118°/1mm Beilstein Reference: 2042374 MDL No.: MFCD00003366
- CAS Number:7311-34-4
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00003366
- Molecular Weight:166.18 g/mol
- Boiling Pt:115…118 °C (1 mmHg)
- Melting Pt:44…48 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:230-772-6
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



