Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ethyl propiolate ≥99%

Glass bottle for solids, 25 g
Catalog # A15068.14
Flammable
Irritant
Ethyl propiolate ≥99%
Catalog # A15068.14
CAS Number:  623-47-2
Ethyl propiolate ≥99%
Ethyl propiolate ≥99%

Glass bottle for solids, 25 g

CAS Number:  623-47-2
Price
164,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 623-47-2 Molecular Formula: C5H6O2 Formula Weight: 98.10 Storage Temperature: 2°C to 8°C Flash Point: 23°(73°F) Refractive Index: 1.4120 Odour: Pungent Melting Point: 9° Boiling Point: 118-120° Beilstein Reference: 878250 UN No.: 3272 MDL No.: MFCD00009184

Synonyms:  Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester
  • CAS Number:623-47-2
  • Formula:C₅H₆O₂
  • MDL Number:MFCD00009184
  • Molecular Weight:98.1 g/mol
  • Boiling Pt:118…120 °C
  • Melting Pt:9 °C
  • Density:0.963
  • Flash Pt:23 °C (73 °F)
  • EINECS:210-795-8
  • UN:3272
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications