Order Entry
Export
Orders LinkContactUsLinkComponent
 

n-Dodecane ≥99%

Glass bottle, 500 mL
Catalog # A14834.AP
Health
n-Dodecane ≥99%
Catalog # A14834.AP
CAS Number:  112-40-3
n-Dodecane ≥99%
n-Dodecane ≥99%

Glass bottle, 500 mL

CAS Number:  112-40-3
Price
140,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99+% CAS No.: 112-40-3 Molecular Formula: CH3(CH2)10CH3 Formula Weight: 170.34 Flash Point: 71°(159°F) Physical Form: Liquid Density: 0.749 Refractive Index: 1.4220 Odour: Petrol-like Melting Point: -10° Boiling Point: 215-217° Beilstein Reference: 1697175 MDL No.: MFCD00008969

Synonyms:  Dodecane
  • CAS Number:112-40-3
  • Formula:H₃C(CH₂)₁₀CH₃
  • MDL Number:MFCD00008969
  • Molecular Weight:170.34 g/mol
  • Boiling Pt:215…217 °C
  • Melting Pt:–10 °C
  • Density:0.749
  • Flash Pt:71 °C (159 °F)
  • Storage Temperature:Ambient
  • EINECS:203-967-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications