Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tris(4-fluorophenyl)phosphine ≥98%

Glass bottle for solids, 25 g
Catalog # A14813.14
Irritant
Tris(4-fluorophenyl)phosphine ≥98%
Catalog # A14813.14
CAS Number:  18437-78-0
Tris(4-fluorophenyl)phosphine ≥98%
Tris(4-fluorophenyl)phosphine ≥98%

Glass bottle for solids, 25 g

CAS Number:  18437-78-0
Price
535,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 18437-78-0 Molecular Formula: C18H12F3P Formula Weight: 316.27 Melting Point: 78-82° Beilstein Reference: 919838

  • CAS Number:18437-78-0
  • Formula:C₁₈H₁₂F₃P
  • MDL Number:MFCD00013553
  • Molecular Weight:316.26 g/mol
  • Melting Pt:78…82 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications