Capric acid ≥99%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 334-48-5 Molecular Formula: CH3(CH2)8CO2H Formula Weight: 172.27 Flash Point: 147°(296°F) Physical Form: Cryst solid Density: 0.893 Appearance: White pH: 4 Melting Point: 29-33° Boiling Point: 268-270° Beilstein Reference: 1754556 Merck Reference: 14,1758 MDL No.: MFCD00004441
Synonyms: Decylic acid, Decanoic acid
- CAS Number:334-48-5
- Formula:C₁₀H₂₀O₂
- MDL Number:MFCD00004441
- Molecular Weight:172.27 g/mol
- Merck Index:14,1758
- Boiling Pt:268…270 °C
- Melting Pt:29…33 °C
- Density:0.893
- Flash Pt:147 °C (296 °F)
- Storage Temperature:Ambient
- EINECS:206-376-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g



