Order Entry
Export
Orders LinkContactUsLinkComponent
 

8-Quinolinol ≥99%

Glass bottle for solids, 100 g
Catalog # A14720.22
Health
Irritant
8-Quinolinol ≥99%
Catalog # A14720.22
CAS Number:  148-24-3
8-Quinolinol ≥99%
8-Quinolinol ≥99%

Glass bottle for solids, 100 g

CAS Number:  148-24-3
Price
51,80 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 148-24-3 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 72-76° Boiling Point: 267° Solubility: Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene. Beilstein Reference: 114512 Merck Reference: 14,4843 MDL No.: MFCD00006807

Synonyms:  8-Hydroxyquinoline
  • CAS Number:148-24-3
  • Formula:C₉H₇NO
  • MDL Number:MFCD00006807
  • Molecular Weight:145.16 g/mol
  • Merck Index:14,4843
  • Boiling Pt:267 °C
  • Melting Pt:72…76 °C
  • Density:1.03 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-711-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications