Order Entry
Export
Orders LinkContactUsLinkComponent
Tricine ≥98%

Tricine ≥98%

Glass bottle for solids, 500 g
Catalog # A14695.36
Tricine ≥98%
Catalog # A14695.36
CAS Number:  5704-04-1
Tricine ≥98%
Tricine ≥98%

Glass bottle for solids, 500 g

CAS Number:  5704-04-1
Price
278,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 5704-04-1 Molecular Formula: (HOCH2)3CNHCH2CO2H Formula Weight: 179.17 Melting Point: ca 183° dec. Beilstein Reference: 1937804 Merck Reference: 14,9651 MDL No.: MFCD00004277

Synonyms:  2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
  • CAS Number:5704-04-1
  • Formula:(HOCH₂)₃CNHCH₂CO₂H
  • MDL Number:MFCD00004277
  • Molecular Weight:179.17 g/mol
  • Merck Index:14,9651
  • Melting Pt:∼ 183 °C (dec.)
  • Density:1.523 g/cm³ (19.85 °C)
  • Storage Temperature:Ambient
  • EINECS:227-193-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications