Tricine ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 5704-04-1 Molecular Formula: (HOCH2)3CNHCH2CO2H Formula Weight: 179.17 Melting Point: ca 183° dec. Beilstein Reference: 1937804 Merck Reference: 14,9651 MDL No.: MFCD00004277
Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
- CAS Number:5704-04-1
- Formula:(HOCH₂)₃CNHCH₂CO₂H
- MDL Number:MFCD00004277
- Molecular Weight:179.17 g/mol
- Merck Index:14,9651
- Melting Pt:∼ 183 °C (dec.)
- Density:1.523 g/cm³ (19.85 °C)
- Storage Temperature:Ambient
- EINECS:227-193-6
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



