Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sarcosine ≥98%

Glass bottle for solids, 100 g
Catalog # A14594.22
Sarcosine ≥98%
Catalog # A14594.22
CAS Number:  107-97-1
Sarcosine ≥98%
Sarcosine ≥98%

Glass bottle for solids, 100 g

CAS Number:  107-97-1
Price
34,10 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 107-97-1 Molecular Formula: CH3NHCH2CO2H Formula Weight: 89.09 Odour: Odourless pH: 6.1 Melting Point: ca 208° dec. Beilstein Reference: 1699442 Merck Reference: 14,8373 MDL No.: MFCD00004279

Synonyms:  H-Sar-OH, N-Methylglycine, Methylaminoacetic acid
  • CAS Number:107-97-1
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00004279
  • Molecular Weight:89.09 g/mol
  • Merck Index:14,8373
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:∼ 208 °C (dec.)
  • Density:1.301 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-538-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications