Order Entry
Export
Orders LinkContactUsLinkComponent
2-Hydroxypyridine ≥98%

2-Hydroxypyridine ≥98%

Glass bottle for solids, 25 g
Catalog # A14522.14
Toxic
Irritant
2-Hydroxypyridine ≥98%
Catalog # A14522.14
CAS Number:  142-08-5
2-Hydroxypyridine ≥98%
2-Hydroxypyridine ≥98%

Glass bottle for solids, 25 g

CAS Number:  142-08-5
Price
36,80 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 142-08-5 Molecular Formula: C5H5NO Formula Weight: 95.10 Storage Temperature: 2°C to 8°C Flash Point: 210°(410°F) Odour: Recognizable Melting Point: 102-111° Boiling Point: 280-281° Beilstein Reference: 105786 UN No.: 2811 MDL No.: MFCD00006268

Synonyms:  Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
  • CAS Number:142-08-5
  • Formula:C₅H₅NO
  • MDL Number:MFCD00006268
  • Molecular Weight:95.10 g/mol
  • Boiling Pt:280…281 °C
  • Melting Pt:102…111 °C
  • Density:1.39
  • Flash Pt:210 °C (410 °F)
  • Storage Temperature:Refrigerator
  • EINECS:205-520-3
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications