Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dimethylbenzonitrile ≥97%

Glass bottle for solids, 1 g
Catalog # A14461.03
Irritant
2,6-Dimethylbenzonitrile ≥97%
Catalog # A14461.03
CAS Number:  6575-13-9
2,6-Dimethylbenzonitrile ≥97%
2,6-Dimethylbenzonitrile ≥97%

Glass bottle for solids, 1 g

CAS Number:  6575-13-9
Price
29,30 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 6575-13-9 Molecular Formula: C9H9N Formula Weight: 131.18 Odour: Almond-like Melting Point: 88-92° Beilstein Reference: 1932523 MDL No.: MFCD00037166

  • CAS Number:6575-13-9
  • Formula:C₉H₉N
  • MDL Number:MFCD00037166
  • Molecular Weight:131.18 g/mol
  • Melting Pt:88…92 °C
  • Storage Temperature:Ambient
  • EINECS:229-503-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications