Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,1-Diphenylethylene ≥98%

Glass bottle for solids, 100 g
Catalog # A14434.22
1,1-Diphenylethylene ≥98%
Catalog # A14434.22
CAS Number:  530-48-3
1,1-Diphenylethylene ≥98%
1,1-Diphenylethylene ≥98%

Glass bottle for solids, 100 g

CAS Number:  530-48-3
Price
356,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 530-48-3 Molecular Formula: C14H12 Formula Weight: 180.25 Flash Point: 136°(276°F) Refractive Index: 1.6065 Melting Point: 5-7° Boiling Point: 270-271° Beilstein Reference: 1099062 Merck Reference: 14,3323 MDL No.: MFCD00008583

Synonyms:  asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
  • CAS Number:530-48-3
  • Formula:(C₆H₅)₂CCH₂
  • MDL Number:MFCD00008583
  • Molecular Weight:180.25 g/mol
  • Merck Index:14,3323
  • Boiling Pt:270…271 °C
  • Melting Pt:5…7 °C
  • Density:1.023
  • Flash Pt:136 °C (276 °F)
  • Storage Temperature:Ambient
  • EINECS:208-482-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications