Orders LinkContactUsLinkComponent
1,1-Diphenylethylene ≥98%
Glass bottle for solids, 25 g
  A14434.14
1,1-Diphenylethylene ≥98%
  A14434.14
 :  530-48-3
1,1-Diphenylethylene ≥98%
1,1-Diphenylethylene ≥98%
Glass bottle for solids, 25 g
 :  530-48-3
118,00 €
1 * 25 g
Purity: 98% CAS No.: 530-48-3 Molecular Formula: C14H12 Formula Weight: 180.25 Flash Point: 136°(276°F) Refractive Index: 1.6065 Melting Point: 5-7° Boiling Point: 270-271° Beilstein Reference: 1099062 Merck Reference: 14,3323 MDL No.: MFCD00008583
 :  asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
  •  :530-48-3
  •  :(C₆H₅)₂CCH₂
  •  :MFCD00008583
  •  :180.25 g/mol
  •  :14,3323
  •  :270…271 °C
  •  :5…7 °C
  •  :1.023
  •  :136 °C (276 °F)
  •  :Ambient
  •  :208-482-6
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g