(±)-Linalool ≥97%
Glass bottle, 2.500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 78-70-6 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 76°(168°F) Physical Form: Liquid Density: 0.865 Refractive Index: 1.4620 Appearance: Colourless Boiling Point: 199-200° Beilstein Reference: 1721488 Merck Reference: 14,5495 MDL No.: MFCD00008906
Synonyms: (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
- CAS Number:78-70-6
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00008906
- Molecular Weight:154.25 g/mol
- Merck Index:14,5495
- Boiling Pt:199…200 °C
- Density:0.865
- Flash Pt:76 °C (168 °F)
- Storage Temperature:Ambient
- EINECS:201-134-4
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2.500 mL



