Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Nitrophenol ≥99%

Glass bottle for solids, 1.000 g
Catalog # A14376.0B
Health
Irritant
4-Nitrophenol ≥99%
Catalog # A14376.0B
CAS Number:  100-02-7
4-Nitrophenol ≥99%
4-Nitrophenol ≥99%

Glass bottle for solids, 1.000 g

CAS Number:  100-02-7
Price
99,40 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 100-02-7 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Flash Point: 169°(336°F) Melting Point: 110-115° Boiling Point: 279° dec. Solubility: Freely soluble in alcohol, chloroform, ether Beilstein Reference: 1281877 Merck Reference: 14,6621 UN No.: 1663 MDL No.: MFCD00007331

Synonyms:  1-Hydroxy-4-nitrobenzene, p-Nitrophenol
  • CAS Number:100-02-7
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD00007331
  • Molecular Weight:139.11 g/mol
  • Merck Index:14,6621
  • Boiling Pt:279 °C (dec.)
  • Melting Pt:110…115 °C
  • Density:1.480
  • Flash Pt:169 °C (336 °F)
  • Storage Temperature:Ambient
  • EINECS:202-811-7
  • UN:1663
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications