Orders LinkContactUsLinkComponent
1,9-Dibromononane ≥97%
1,9-Dibromononane ≥97%
Glass bottle for solids, 25 g
  A14324.14
1,9-Dibromononane ≥97%
  A14324.14
 :  4549-33-1
1,9-Dibromononane ≥97%
1,9-Dibromononane ≥97%
Glass bottle for solids, 25 g
 :  4549-33-1
93,70 €
1 * 25 g
Purity: 97% CAS No.: 4549-33-1 Molecular Formula: C9H18Br2 Formula Weight: 286.06 Flash Point: 110°(230°F) Refractive Index: 1.4965 Boiling Point: 285-288° Beilstein Reference: 1737598 MDL No.: MFCD00000279
 :  Nonamethylene dibromide
  •  :4549-33-1
  •  :BrH₂C(CH₂)₇CH₂Br
  •  :MFCD00000279
  •  :286.05 g/mol
  •  :285…288 °C
  •  :–10 °C
  •  :1.410
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :224-913-0
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g