Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,9-Dibromononane ≥97%

Glass bottle for solids, 10 g
Catalog # A14324.09
Irritant
1,9-Dibromononane ≥97%
Catalog # A14324.09
CAS Number:  4549-33-1
1,9-Dibromononane ≥97%
1,9-Dibromononane ≥97%

Glass bottle for solids, 10 g

CAS Number:  4549-33-1
Price
48,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 97% CAS No.: 4549-33-1 Molecular Formula: C9H18Br2 Formula Weight: 286.06 Flash Point: 110°(230°F) Refractive Index: 1.4965 Boiling Point: 285-288° Beilstein Reference: 1737598 MDL No.: MFCD00000279

Synonyms:  Nonamethylene dibromide
  • CAS Number:4549-33-1
  • Formula:BrH₂C(CH₂)₇CH₂Br
  • MDL Number:MFCD00000279
  • Molecular Weight:286.05 g/mol
  • Boiling Pt:285…288 °C
  • Melting Pt:–10 °C
  • Density:1.410
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:224-913-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Cat. No.:
    A14324.09

Specifications