Order Entry
Export
Orders LinkContactUsLinkComponent
Bathophenanthroline ≥98%

Bathophenanthroline ≥98%

Glass bottle for solids, 25 g
Catalog # A14258.14
Bathophenanthroline ≥98%
Catalog # A14258.14
CAS Number:  1662-01-7
Bathophenanthroline ≥98%
Bathophenanthroline ≥98%

Glass bottle for solids, 25 g

CAS Number:  1662-01-7
Price
1 980,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 1662-01-7 Molecular Formula: C24H16N2 Formula Weight: 332.41 Melting Point: 218-222° Beilstein Reference: 261048

Synonyms:  4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
  • CAS Number:1662-01-7
  • Formula:C₂₄H₁₆N₂
  • MDL Number:MFCD00004976
  • Molecular Weight:332.4 g/mol
  • Boiling Pt:508 °C (1013 hPa)
  • Melting Pt:218…222 °C
  • Storage Temperature:Ambient
  • EINECS:216-767-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications