Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bathophenanthroline ≥98%

Glass bottle for solids, 1 g
Bathophenanthroline ≥98%
  A14258.03
 :  1662-01-7
Bathophenanthroline ≥98%
Bathophenanthroline ≥98%

Glass bottle for solids, 1 g

 :  1662-01-7
 
107,00 €
Log in for availability
1 * 1 g
 

Purity: 98+% CAS No.: 1662-01-7 Molecular Formula: C24H16N2 Formula Weight: 332.41 Melting Point: 218-222° Beilstein Reference: 261048

Synonyms:  4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
  • CAS Number:1662-01-7
  • Formula:C₂₄H₁₆N₂
  • MDL Number:MFCD00004976
  • Molecular Weight:332.4 g/mol
  • Boiling Pt:508 °C (1013 hPa)
  • Melting Pt:218…222 °C
  • Storage Temperature:Ambient
  • EINECS:216-767-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications