Order Entry
Export
Orders LinkContactUsLinkComponent
3-Fluoroaniline ≥98%

3-Fluoroaniline ≥98%

Glass bottle for solids, 100 g
Catalog # A14228.22
Toxic
Health
Irritant
3-Fluoroaniline ≥98%
Catalog # A14228.22
CAS Number:  372-19-0
3-Fluoroaniline ≥98%
3-Fluoroaniline ≥98%

Glass bottle for solids, 100 g

CAS Number:  372-19-0
Price
186,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 372-19-0 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 77°(170°F) Refractive Index: 1.5440 Odour: Recognizable Boiling Point: 186-187° Beilstein Reference: 1305471 UN No.: 2941 MDL No.: MFCD00007758

  • CAS Number:372-19-0
  • Formula:C₆H₆FN
  • MDL Number:MFCD00007758
  • Molecular Weight:111.12 g/mol
  • Boiling Pt:186…187 °C
  • Melting Pt:–2 °C
  • Density:1.156
  • Flash Pt:77 °C (170 °F)
  • Storage Temperature:Ambient
  • EINECS:206-747-0
  • UN:2941
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications