Order Entry
Export
Orders LinkContactUsLinkComponent
Sebacoyl dichloride ≥97%

Sebacoyl dichloride ≥97%

Glass bottle for solids, 500 g
Catalog # A14226.36
Toxic
Corrosive
Sebacoyl dichloride ≥97%
Catalog # A14226.36
CAS Number:  111-19-3
Sebacoyl dichloride ≥97%
Sebacoyl dichloride ≥97%

Glass bottle for solids, 500 g

CAS Number:  111-19-3
Price
379,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 111-19-3 Molecular Formula: ClCO(CH2)8COCl Formula Weight: 239.14 Flash Point: 165°(329°F) Refractive Index: 1.4680 Odour: Acrid Melting Point: -5° Boiling Point: 161°/14mm Beilstein Reference: 1365665 UN No.: 2922 MDL No.: MFCD00000770

Synonyms:  Sebacoyl chloride
  • CAS Number:111-19-3
  • Formula:C₁₀H₁₆Cl₂O₂
  • MDL Number:MFCD00000770
  • Molecular Weight:239.14 g/mol
  • Boiling Pt:161 °C (14 mmHg)
  • Melting Pt:–5 °C
  • Density:1.121
  • Flash Pt:165 °C (329 °F)
  • Storage Temperature:Ambient
  • EINECS:203-843-4
  • UN:2922
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications