Orders LinkContactUsLinkComponent
Sebacoyl dichloride ≥97%
Glass bottle for solids, 100 g
  A14226.22
Sebacoyl dichloride ≥97%
  A14226.22
 :  111-19-3
Sebacoyl dichloride ≥97%
Sebacoyl dichloride ≥97%
Glass bottle for solids, 100 g
 :  111-19-3
114,00 €
1 * 100 g
Purity: 97% CAS No.: 111-19-3 Molecular Formula: ClCO(CH2)8COCl Formula Weight: 239.14 Flash Point: 165°(329°F) Refractive Index: 1.4680 Odour: Acrid Melting Point: -5° Boiling Point: 161°/14mm Beilstein Reference: 1365665 UN No.: 2922 MDL No.: MFCD00000770
 :  Sebacoyl chloride
  •  :111-19-3
  •  :C₁₀H₁₆Cl₂O₂
  •  :MFCD00000770
  •  :239.14 g/mol
  •  :161 °C (14 mmHg)
  •  :–5 °C
  •  :1.121
  •  :165 °C (329 °F)
  •  :Ambient
  •  :203-843-4
  •  :2922
  •  :8,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g