Orders LinkContactUsLinkComponent
(±)-Mandelic acid ≥99%
Glass bottle for solids, 2.500 g
  A14223.0E
(±)-Mandelic acid ≥99%
  A14223.0E
 :  90-64-2
(±)-Mandelic acid ≥99%
(±)-Mandelic acid ≥99%
Glass bottle for solids, 2.500 g
 :  90-64-2
334,00 €
1 * 2.500 g
Purity: 99% CAS No.: 90-64-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Physical Form: Cryst powder Sensitivity: Light Sensitive Appearance: White Melting Point: 118-121° Beilstein Reference: 510011 Merck Reference: 14,5717 MDL No.: MFCD00064250
 :  DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
  •  :90-64-2
  •  :C₈H₈O₃
  •  :MFCD00064250
  •  :152.15 g/mol
  •  :14,5717
  •  :322 °C (1013 hPa)
  •  :118…121 °C
  •  :1.361 g/cm³ (20 °C)
  •  :Ambient
  •  :202-007-6
  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g