Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Benzodioxole ≥99%

1,3-Benzodioxole ≥99%

Glass bottle for solids, 1.000 g
Catalog # A14192.0B
Irritant
1,3-Benzodioxole ≥99%
Catalog # A14192.0B
CAS Number:  274-09-9
1,3-Benzodioxole ≥99%
1,3-Benzodioxole ≥99%

Glass bottle for solids, 1.000 g

CAS Number:  274-09-9
Price
733,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 274-09-9 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 55°(131°F) Refractive Index: 1.5390 Odour: Aromatic Melting Point: -18° Boiling Point: 172-173° Beilstein Reference: 115506 MDL No.: MFCD00005818

Synonyms:  Benzo[d][1,3]dioxole, 1,2-Methylenedioxybenzene
  • CAS Number:274-09-9
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00005818
  • Molecular Weight:122.12 g/mol
  • Boiling Pt:172…173 °C
  • Melting Pt:–18 °C
  • Density:1.185
  • Flash Pt:55 °C (131 °F)
  • Storage Temperature:Ambient
  • EINECS:205-992-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications