Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dioxane ≥99% stabilised

1,4-Dioxane ≥99% stabilised

Glass bottle for solids, 500 g
Catalog # A14188.36
Flammable
Health
Irritant
1,4-Dioxane ≥99% stabilised
Catalog # A14188.36
CAS Number:  123-91-1
1,4-Dioxane ≥99% stabilised
1,4-Dioxane ≥99% stabilised

Glass bottle for solids, 500 g

CAS Number:  123-91-1
Price
43,20 €
Log in for availability
1 * 500 g
Product Details & Documents

Stabilized with ca 5-10 ppm BHT

Purity: 99+% CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Refractive Index: 1.4220 Odour: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571

Synonyms:  Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
  • CAS Number:123-91-1
  • Formula:C₄H₈O₂
  • MDL Number:MFCD00006571
  • Molecular Weight:88.11 g/mol
  • Merck Index:14,3300
  • Boiling Pt:100…102 °C
  • Melting Pt:11.8 °C
  • Density:1.034
  • Flash Pt:12 °C (54 °F)
  • Storage Temperature:Ambient
  • UN:1165
  • ADR:3,II
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 5-10 ppm
  • EINECS:204-661-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications