Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dichloroacetone 96%

Glass bottle for solids, 500 g
Catalog # A14149.36
Toxic
Corrosive
Health
1,3-Dichloroacetone 96%
Catalog # A14149.36
CAS Number:  534-07-6
1,3-Dichloroacetone 96%
1,3-Dichloroacetone 96%

Glass bottle for solids, 500 g

CAS Number:  534-07-6
Price
522,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 96% CAS No.: 534-07-6 Molecular Formula: C3H4Cl2O Formula Weight: 126.97 Storage Temperature: 2°C to 8°C Flash Point: 95°(203°F) Refractive Index: 1.4400 Odour: Pungent pH: 4.2 Melting Point: 40-44° Boiling Point: 172-173° Beilstein Reference: 605456 Merck Reference: 14,3052 UN No.: 2649 MDL No.: MFCD00000937

  • CAS Number:534-07-6
  • Formula:ClH₂CC(O)CH₂Cl
  • MDL Number:MFCD00000937
  • Molecular Weight:126.97 g/mol
  • Merck Index:14,3052
  • Boiling Pt:172…173 °C
  • Melting Pt:40…44 °C
  • Density:1.383
  • Flash Pt:95 °C (203 °F)
  • Storage Temperature:Refrigerator
  • EINECS:208-585-6
  • UN:2649
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications