Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Difluorobenzonitrile ≥98%

2,4-Difluorobenzonitrile ≥98%

Glass bottle for solids, 25 g
Catalog # A14113.14
Toxic
Irritant
2,4-Difluorobenzonitrile ≥98%
Catalog # A14113.14
CAS Number:  3939-09-1
2,4-Difluorobenzonitrile ≥98%
2,4-Difluorobenzonitrile ≥98%

Glass bottle for solids, 25 g

CAS Number:  3939-09-1
Price
137,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 3939-09-1 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 110°(230°F) Odour: Almond-like Boiling Point: 189° Beilstein Reference: 1940316 UN No.: 3439 MDL No.: MFCD00009826

  • CAS Number:3939-09-1
  • Formula:C₇H₃F₂N
  • MDL Number:MFCD00009826
  • Molecular Weight:139.1 g/mol
  • Boiling Pt:189 °C
  • Melting Pt:43…48 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:223-523-8
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications