Orders LinkContactUsLinkComponent
2-Mercaptobenzothiazole ≥97%
2-Mercaptobenzothiazole ≥97%
Glass bottle for solids, 500 g
  A14086.36
2-Mercaptobenzothiazole ≥97%
  A14086.36
 :  149-30-4
2-Mercaptobenzothiazole ≥97%
2-Mercaptobenzothiazole ≥97%
Glass bottle for solids, 500 g
 :  149-30-4
50,10 €
1 * 500 g
Purity: 97% CAS No.: 149-30-4 Molecular Formula: C7H5NS2 Formula Weight: 167.25 Flash Point: 243°(469°F) Melting Point: 178-182° Beilstein Reference: 119484 Merck Reference: 14,5868 UN No.: 3077 MDL No.: MFCD00005781
 :  2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
  •  :149-30-4
  •  :C₇H₅NS₂
  •  :MFCD00005781
  •  :167.26 g/mol
  •  :14,5868
  •  :302 °C (1013 hPa)
  •  :178…182 °C
  •  :1.420
  •  :243 °C (469 °F)
  •  :Ambient
  •  :205-736-8
  •  :3077
  •  :9,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g