Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Mercaptobenzothiazole ≥97%

Glass bottle for solids, 10 g
Catalog # A14086.09
Irritant
Environment
2-Mercaptobenzothiazole ≥97%
Catalog # A14086.09
CAS Number:  149-30-4
2-Mercaptobenzothiazole ≥97%
2-Mercaptobenzothiazole ≥97%

Glass bottle for solids, 10 g

CAS Number:  149-30-4
Price
22,30 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 97% CAS No.: 149-30-4 Molecular Formula: C7H5NS2 Formula Weight: 167.25 Flash Point: 243°(469°F) Melting Point: 178-182° Beilstein Reference: 119484 Merck Reference: 14,5868 UN No.: 3077 MDL No.: MFCD00005781

Synonyms:  2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
  • CAS Number:149-30-4
  • Formula:C₇H₅NS₂
  • MDL Number:MFCD00005781
  • Molecular Weight:167.26 g/mol
  • Merck Index:14,5868
  • Boiling Pt:302 °C (1013 hPa)
  • Melting Pt:178…182 °C
  • Density:1.420
  • Flash Pt:243 °C (469 °F)
  • Storage Temperature:Ambient
  • EINECS:205-736-8
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications