Order Entry
Export
Orders LinkContactUsLinkComponent
Benzoic acid ≥99%

Benzoic acid ≥99%

Glass bottle for solids, 250 g
Catalog # A14062.30
Corrosive
Irritant
Benzoic acid ≥99%
Catalog # A14062.30
CAS Number:  65-85-0
Benzoic acid ≥99%
Benzoic acid ≥99%

Glass bottle for solids, 250 g

CAS Number:  65-85-0
Price
27,50 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 65-85-0 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 121°(250°F) Physical Form: Crystalline powder Density: 1.32 Appearance: White Odour: Weak, characteristic pH: 2.5-3.5 Melting Point: 121-124° Boiling Point: 249° Beilstein Reference: 636131 Merck Reference: 14,1091 MDL No.: MFCD00002398

  • CAS Number:65-85-0
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00002398
  • Molecular Weight:122.12 g/mol
  • Merck Index:14,1091
  • Boiling Pt:249 °C
  • Melting Pt:121…124 °C
  • Density:1.32
  • Flash Pt:121 °C (250 °F)
  • Storage Temperature:Ambient
  • EINECS:200-618-2
  • REACH:01-2119455536-33
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications