Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Difluorophenol ≥98%

Glass bottle for solids, 250 g
Catalog # A14044.30
Toxic
Corrosive
Flammable
2,4-Difluorophenol ≥98%
Catalog # A14044.30
CAS Number:  367-27-1
2,4-Difluorophenol ≥98%
2,4-Difluorophenol ≥98%

Glass bottle for solids, 250 g

CAS Number:  367-27-1
Price
565,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98+% CAS No.: 367-27-1 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 56°(132°F) Refractive Index: 1.4860 Odour: Phenol-like Melting Point: 21-24° Boiling Point: 52-54°/14mm Beilstein Reference: 1934803 UN No.: 2921 MDL No.: MFCD00009715

  • CAS Number:367-27-1
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00009715
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:52…54 °C (14 mmHg)
  • Melting Pt:21…24 °C
  • Density:1.362
  • Flash Pt:56 °C (132 °F)
  • Storage Temperature:Ambient
  • EINECS:206-688-0
  • UN:2921
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications