2,4-Difluorophenol ≥98%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 367-27-1 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 56°(132°F) Refractive Index: 1.4860 Odour: Phenol-like Melting Point: 21-24° Boiling Point: 52-54°/14mm Beilstein Reference: 1934803 UN No.: 2921 MDL No.: MFCD00009715
- CAS Number:367-27-1
- Formula:C₆H₄F₂O
- MDL Number:MFCD00009715
- Molecular Weight:130.09 g/mol
- Boiling Pt:52…54 °C (14 mmHg)
- Melting Pt:21…24 °C
- Density:1.362
- Flash Pt:56 °C (132 °F)
- Storage Temperature:Ambient
- EINECS:206-688-0
- UN:2921
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



