Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(-)-Mannitol ≥97.0%, white powder

Glass bottle for solids, 5.000 g
Catalog # A14030.0I
D-(-)-Mannitol ≥97.0%, white powder
Catalog # A14030.0I
CAS Number:  69-65-8
D-(-)-Mannitol ≥97.0%, white powder
D-(-)-Mannitol ≥97.0%, white powder

Glass bottle for solids, 5.000 g

CAS Number:  69-65-8
Price
330,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 69-65-8 Molecular Formula: C6H14O6 Formula Weight: 182.17 Odour: Odourless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Beilstein Reference: 1721898 Merck Reference: 14,5745 Optical Rotation: +24° (in borax solution) MDL No.: MFCD00064287

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:14,5745
  • Boiling Pt:290…295 °C (3.5 mmHg)
  • Melting Pt:165…167 °C
  • Density:1.489
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications