Order Entry
Export
Orders LinkContactUsLinkComponent
1,5-Pentanediol ≥97%

1,5-Pentanediol ≥97%

Glass bottle for solids, 500 g
Catalog # A13998.36
Irritant
1,5-Pentanediol ≥97%
Catalog # A13998.36
CAS Number:  111-29-5
1,5-Pentanediol ≥97%
1,5-Pentanediol ≥97%

Glass bottle for solids, 500 g

CAS Number:  111-29-5
Price
64,60 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 111-29-5 Molecular Formula: C5H12O2 Formula Weight: 104.15 Flash Point: 129°(264°F) Refractive Index: 1.4500 Melting Point: -16° Boiling Point: 241-242° Beilstein Reference: 1560130 Merck Reference: 14,7117 MDL No.: MFCD00002978

Synonyms:  Pentane-1,5-diol, 1,5-Dihydroxy pentane
  • CAS Number:111-29-5
  • Formula:C₅H₁₂O₂
  • MDL Number:MFCD00002978
  • Molecular Weight:104.15 g/mol
  • Merck Index:14,7117
  • Boiling Pt:241…242 °C
  • Melting Pt:–16 °C
  • Density:0.992
  • Flash Pt:129 °C (264 °F)
  • Storage Temperature:Ambient
  • EINECS:203-854-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications