Order Entry
Export
Orders LinkContactUsLinkComponent
1,5-Dinitronaphthalene ≥97%

1,5-Dinitronaphthalene ≥97%

Glass bottle for solids, 100 g
Catalog # A13869.22
Health
Irritant
1,5-Dinitronaphthalene ≥97%
Catalog # A13869.22
CAS Number:  605-71-0
1,5-Dinitronaphthalene ≥97%
1,5-Dinitronaphthalene ≥97%

Glass bottle for solids, 100 g

CAS Number:  605-71-0
Price
54,30 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97+% CAS No.: 605-71-0 Molecular Formula: C10H6N2O4 Formula Weight: 218.17 Odour: Odourless Melting Point: 214-218° Beilstein Reference: 527184 MDL No.: MFCD00003916

  • CAS Number:605-71-0
  • Formula:C₁₀H₆N₂O₄
  • MDL Number:MFCD00003916
  • Molecular Weight:218.17 g/mol
  • Boiling Pt:390 °C (1013 hPa)
  • Melting Pt:214…218 °C
  • Density:1.586
  • Storage Temperature:Ambient
  • EINECS:210-095-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications