Triiodomethane ≥99%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 75-47-8 Molecular Formula: CHI3 Formula Weight: 393.73 Flash Point: None Odour: Unpleasant Beilstein Reference: 1697010 Merck Reference: 14,5033 MDL No.: MFCD00001069
Synonyms: Iodoform
- CAS Number:75-47-8
- Formula:CHI₃
- MDL Number:MFCD00001069
- Molecular Weight:393.73 g/mol
- Merck Index:14,5033
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:118…123 °C
- Density:4.008
- Storage Temperature:Ambient
- EINECS:200-874-5
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



