Order Entry
Export
Orders LinkContactUsLinkComponent
Triiodomethane ≥99%

Triiodomethane ≥99%

Glass bottle for solids, 1.000 g
Catalog # A13664.0B
Irritant
Triiodomethane ≥99%
Catalog # A13664.0B
CAS Number:  75-47-8
Triiodomethane ≥99%
Triiodomethane ≥99%

Glass bottle for solids, 1.000 g

CAS Number:  75-47-8
Price
452,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 75-47-8 Molecular Formula: CHI3 Formula Weight: 393.73 Flash Point: None Odour: Unpleasant Beilstein Reference: 1697010 Merck Reference: 14,5033 MDL No.: MFCD00001069

Synonyms:  Iodoform
  • CAS Number:75-47-8
  • Formula:CHI₃
  • MDL Number:MFCD00001069
  • Molecular Weight:393.73 g/mol
  • Merck Index:14,5033
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:118…123 °C
  • Density:4.008
  • Storage Temperature:Ambient
  • EINECS:200-874-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications