Order Entry
Export
Orders LinkContactUsLinkComponent
1,5-Hexadiene ≥98%

1,5-Hexadiene ≥98%

Glass bottle for solids, 100 g
Catalog # A13650.22
Flammable
Health
Irritant
1,5-Hexadiene ≥98%
Catalog # A13650.22
CAS Number:  592-42-7
1,5-Hexadiene ≥98%
1,5-Hexadiene ≥98%

Glass bottle for solids, 100 g

CAS Number:  592-42-7
Price
107,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 592-42-7 Molecular Formula: CH2=CHCH2CH2CH=CH2 Formula Weight: 82.15 Flash Point: -27°(-16°F) Refractive Index: 1.4040 Odour: Turpentine-like Melting Point: -141° Boiling Point: 58-60° Beilstein Reference: 1731106 UN No.: 2458 MDL No.: MFCD00008666

Synonyms:  Hexa-1,5-diene
  • CAS Number:592-42-7
  • Formula:C₆H₁₀
  • MDL Number:MFCD00008666
  • Molecular Weight:82.15 g/mol
  • Boiling Pt:58…60 °C
  • Melting Pt:–141 °C
  • Density:0.692
  • Flash Pt:–27 °C (–16 °F)
  • Storage Temperature:Ambient
  • EINECS:209-754-7
  • UN:2458
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications