Order Entry
Export
Orders LinkContactUsLinkComponent
2-Ethoxyethyl acetate ≥98%

2-Ethoxyethyl acetate ≥98%

Glass bottle, 500 mL
Catalog # A13639.AP
Health
Flammable
Irritant
2-Ethoxyethyl acetate ≥98%
Catalog # A13639.AP
CAS Number:  111-15-9
2-Ethoxyethyl acetate ≥98%
2-Ethoxyethyl acetate ≥98%

Glass bottle, 500 mL

CAS Number:  111-15-9
Price
31,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 98+% CAS No.: 111-15-9 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 52°(126°F) Refractive Index: 1.4070 Odour: Fruit-like Melting Point: -62° Boiling Point: 155-157° Beilstein Reference: 1748677 Merck Reference: 14,3751 UN No.: 1172 MDL No.: MFCD00009251

Synonyms:  Ethylglycol acetate
  • CAS Number:111-15-9
  • Formula:C₆H₁₂O₃
  • MDL Number:MFCD00009251
  • Molecular Weight:132.16 g/mol
  • Merck Index:14,3751
  • Boiling Pt:155…157 °C
  • Melting Pt:–62 °C
  • Density:0.975
  • Flash Pt:52 °C (126 °F)
  • Storage Temperature:Ambient
  • EINECS:203-839-2
  • UN:1172
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications