Rhodamine B
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 81-88-9 Molecular Formula: C28H31ClN2O3 Formula Weight: 479.02 Physical Form: Crystals Density: 1.31 Appearance: Purple powder/dark green Melting Point: ca 210° dec. Beilstein Reference: 4119648 Merck Reference: 14,8183 MDL No.: MFCD00011931
Suitable for fluorescent labelling
Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
- CAS Number:81-88-9
- Formula:C₂₈H₃₁ClN₂O₃
- MDL Number:MFCD00011931
- Molecular Weight:479.02 g/mol
- Merck Index:14,8183
- Melting Pt:∼ 210 °C (dec.)
- Density:1.31
- Storage Temperature:Ambient
- EINECS:201-383-9
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



