Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Dihydroxybenzaldehyde ≥97%

Glass bottle for solids, 100 g
Catalog # A13490.22
Irritant
2,3-Dihydroxybenzaldehyde ≥97%
Catalog # A13490.22
CAS Number:  24677-78-9
2,3-Dihydroxybenzaldehyde ≥97%
2,3-Dihydroxybenzaldehyde ≥97%

Glass bottle for solids, 100 g

CAS Number:  24677-78-9
Price
462,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 24677-78-9 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 106-109° Beilstein Reference: 2041781

  • CAS Number:24677-78-9
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00003324
  • Molecular Weight:138.12 g/mol
  • Boiling Pt:235 °C (1013 hPa)
  • Melting Pt:106…109 °C
  • Storage Temperature:Ambient
  • EINECS:246-398-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications