Orders LinkContactUsLinkComponent
1,6-Dibromohexane ≥97%
Glass bottle for solids, 500 g
  A13417.36
1,6-Dibromohexane ≥97%
  A13417.36
 :  629-03-8
1,6-Dibromohexane ≥97%
1,6-Dibromohexane ≥97%
Glass bottle for solids, 500 g
 :  629-03-8
91,80 €
1 * 500 g
Purity: 97+% CAS No.: 629-03-8 Molecular Formula: Br(CH2)6Br Formula Weight: 243.98 Flash Point: 110°(230°F) Refractive Index: 1.5075 Odour: Odourless Melting Point: -2° Boiling Point: 242-244° Beilstein Reference: 1236322 UN No.: 3082 MDL No.: MFCD00000272
 :  Hexamethylen dibromide
  •  :629-03-8
  •  :BrH₂C(CH₂)₄CH₂Br
  •  :MFCD00000272
  •  :243.97 g/mol
  •  :242…244 °C
  •  :–2 °C
  •  :1.610
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :211-067-2
  •  :3082
  •  :9,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g