Order Entry
Export
Orders LinkContactUsLinkComponent
1,6-Dibromohexane ≥97%

1,6-Dibromohexane ≥97%

Glass bottle for solids, 100 g
Catalog # A13417.22
Irritant
1,6-Dibromohexane ≥97%
Catalog # A13417.22
CAS Number:  629-03-8
1,6-Dibromohexane ≥97%
1,6-Dibromohexane ≥97%

Glass bottle for solids, 100 g

CAS Number:  629-03-8
Price
31,50 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97+% CAS No.: 629-03-8 Molecular Formula: Br(CH2)6Br Formula Weight: 243.98 Flash Point: 110°(230°F) Refractive Index: 1.5075 Odour: Odourless Melting Point: -2° Boiling Point: 242-244° Beilstein Reference: 1236322 UN No.: 3082 MDL No.: MFCD00000272

Synonyms:  Hexamethylen dibromide
  • CAS Number:629-03-8
  • Formula:BrH₂C(CH₂)₄CH₂Br
  • MDL Number:MFCD00000272
  • Molecular Weight:243.97 g/mol
  • Boiling Pt:242…244 °C
  • Melting Pt:–2 °C
  • Density:1.610
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:211-067-2
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications