Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Mercaptopyrimidine ≥98%

Glass bottle for solids, 100 g
Catalog # A13382.22
Irritant
2-Mercaptopyrimidine ≥98%
Catalog # A13382.22
CAS Number:  1450-85-7
2-Mercaptopyrimidine ≥98%
2-Mercaptopyrimidine ≥98%

Glass bottle for solids, 100 g

CAS Number:  1450-85-7
Price
336,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 1450-85-7 Molecular Formula: C4H4N2S Formula Weight: 112.15 Melting Point: ca 230° dec. Beilstein Reference: 107105

Synonyms:  2-Thiopyrimidine
  • CAS Number:1450-85-7
  • Formula:C₄H₄N₂S
  • MDL Number:MFCD00006073
  • Molecular Weight:112.16 g/mol
  • Melting Pt:∼ 230 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:215-917-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications