Order Entry
Export
Orders LinkContactUsLinkComponent
Uric acid ≥99%

Uric acid ≥99%

Glass bottle for solids, 500 g
Catalog # A13346.36
Uric acid ≥99%
Catalog # A13346.36
CAS Number:  69-93-2
Uric acid ≥99%
Uric acid ≥99%

Glass bottle for solids, 500 g

CAS Number:  69-93-2
Price
647,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 69-93-2 Molecular Formula: C5H4N4O3 Formula Weight: 168.11 Melting Point: 300° Beilstein Reference: 156158 Merck Reference: 14,9875 MDL No.: MFCD00005712

Synonyms:  1H-Purine-2,6,8(3H,7H,9H)-trione
  • CAS Number:69-93-2
  • Formula:C₅H₄N₄O₃
  • MDL Number:MFCD00005712
  • Molecular Weight:168.11 g/mol
  • Merck Index:14,9875
  • Melting Pt:>300 °C
  • Density:1.89
  • Storage Temperature:Ambient
  • EINECS:200-720-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications