Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Bromopyridine ≥99%

Glass bottle for solids, 250 g
Catalog # A13241.30
Toxic
Irritant
2-Bromopyridine ≥99%
Catalog # A13241.30
CAS Number:  109-04-6
2-Bromopyridine ≥99%
2-Bromopyridine ≥99%

Glass bottle for solids, 250 g

CAS Number:  109-04-6
Price
194,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 109-04-6 Molecular Formula: C5H4BrN Formula Weight: 158.00 Flash Point: 97°(207°F) Refractive Index: 1.5720 Odour: Characteristic Boiling Point: 192-194° Beilstein Reference: 105789 UN No.: 2810 MDL No.: MFCD00006219

  • CAS Number:109-04-6
  • Formula:C₅H₄BrN
  • MDL Number:MFCD00006219
  • Molecular Weight:158 g/mol
  • Boiling Pt:192…194 °C
  • Melting Pt:–40.1 °C
  • Density:1.625
  • Flash Pt:97 °C (207 °F)
  • Storage Temperature:Ambient
  • EINECS:203-641-6
  • UN:2810
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications